CAS RN 66564-14-5
CAS RN 66564-14-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Cinitapride, TRC
CAS: 66564-14-5 Molecular Formula: C21 H30 N4 O4 Molecular Weight (g/mol): 402.49 Synonym: 4-Amino-N-[1-(3-cyclohexen-1-ylmethyl)-4-piperidinyl]-2-ethoxy-5-nitrobenzamide,Cinitapride IUPAC Name: 4-amino-N-[1-(cyclohex-3-en-1-ylmethyl)piperidin-4-yl]-2-ethoxy-5-nitrobenzamide SMILES: CCOc1cc(N)c(cc1C(=O)NC2CCN(CC3CCC=CC3)CC2)[N+](=O)[O-]