accessibility menu, dialog, popup

UserName

NH2ClClNOO

CAS RN 6641-64-1

IUPAC Name: 4,5-dichloro-2-nitroaniline
Synonyms: 4,5-dichloro-2-nitroaniline
Molecular Weight (g/mol): 207.01
Molecular Formula: C6H4Cl2N2O2
InChi Key: FSGTULQLEVAYRS-UHFFFAOYSA-N
SMILES: NC1=CC(Cl)=C(Cl)C=C1[N+]([O-])=O
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results