accessibility menu, dialog, popup

UserName

CAS RN 66357-59-3

IUPAC Name: hydrogen dimethyl[(5-{[(2-{[(1E)-1-(methylamino)-2-nitroethenyl]amino}ethyl)sulfanyl]methyl}furan-2-yl)methyl]amine chloride
Synonyms: raniβ ranitidine hcl ranitidine hydrochloride nu-ranit rani-nerton zantac raniberl kuracid ranibloc alquen
Molecular Weight (g/mol): 350.86
Molecular Formula: C13H23ClN4O3S
InChi Key: GGWBHVILAJZWKJ-KJEVSKRMSA-N
SMILES: [H+].[Cl-].CN\C(NCCSCC1=CC=C(CN(C)C)O1)=C/[N+]([O-])=O
Molecular Weight (g/mol) 
  • (8)
  • (2)
Percent Purity 
  • (3)
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
Certifications/Compliance 
  • (2)
  • (1)
Color 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results