accessibility menu, dialog, popup

UserName

CAS RN 6627-89-0

H3CH3CCH3OOO

CAS RN 6627-89-0

IUPAC Name: tert-butyl phenyl carbonate
Synonyms: tert-butyl phenyl carbonate
Molecular Weight (g/mol): 194.23
Molecular Formula: C11H14O3
InChi Key: UXWVQHXKKOGTSY-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)OC1=CC=CC=C1
Quantity 
  • (3)
  • (3)
  • (1)
Boiling Point 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (3)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)