CAS RN 6627-88-9
CAS RN 6627-88-9
IUPAC Name:
2,6-dimethoxy-4-(prop-2-en-1-yl)phenol
Synonyms:
2,6-dimethoxy-4-(prop-2-en-1-yl)phenol
Molecular Weight (g/mol):
194.23
Molecular Formula:
C11H14O3
InChi Key:
FWMPKHMKIJDEMJ-UHFFFAOYSA-N
SMILES:
COC1=CC(CC=C)=CC(OC)=C1O
1
–
1
of
1
results
1
4-Allyl-2,6-dimethoxyphenol, 98%
CAS: 6627-88-9 Molecular Formula: C11H14O3 Molecular Weight (g/mol): 194.23 MDL Number: MFCD00008655 InChI Key: FWMPKHMKIJDEMJ-UHFFFAOYSA-N Synonym: 4-allyl-2,6-dimethoxyphenol,methoxyeugenol,4-allylsyringol,phenol, 2,6-dimethoxy-4-2-propenyl,2,6-dimethoxychavicol,phenol, 4-allyl-2,6-dimethoxy,unii-8vf00ywp89,2,6-dimethoxy-4-allylphenol,2,6-dimethoxy-4-prop-2-en-1-yl phenol,4-hydroxy-3,5-dimethoxyallylbenzene PubChem CID: 226486 ChEBI: CHEBI:86562 IUPAC Name: 2,6-dimethoxy-4-prop-2-enylphenol SMILES: COC1=CC(CC=C)=CC(OC)=C1O