CAS RN 6624-49-3
IUPAC Name:
isoquinoline-3-carboxylic acid
Synonyms:
isoquinoline-3-carboxylic acid
Molecular Weight (g/mol):
173.17
Molecular Formula:
C10H7NO2
InChi Key:
KVMMIDQDXZOPAB-UHFFFAOYSA-N
SMILES:
OC(=O)C1=NC=C2C=CC=CC2=C1
1
–
1
of
1
results
1
Isoquinoline-3-carboxylic acid, 97%, Thermo Scientific™
CAS: 6624-49-3 Molecular Formula: C10H7NO2 Molecular Weight (g/mol): 173.17 InChI Key: KVMMIDQDXZOPAB-UHFFFAOYSA-N Synonym: 3-isoquinolinecarboxylic acid,3-carboxyisoquinoline,3-isoquinoline carboxylic acid,3-isoquinaldic acid,pubchem16889,maybridge1_002411,acmc-1b7v6,isoquinoline-3-caboxylic acid,ksc217c5f,isoquinoline 3-carboxylic acid PubChem CID: 124656 IUPAC Name: isoquinoline-3-carboxylic acid SMILES: C1=CC=C2C=NC(=CC2=C1)C(=O)O