CAS RN 6620-60-6
CAS RN 6620-60-6
IUPAC Name:
4-(dipropylcarbamoyl)-4-(phenylformamido)butanoic acid
Synonyms:
proglumide
Molecular Weight (g/mol):
334.42
Molecular Formula:
C18H26N2O4
InChi Key:
DGMKFQYCZXERLX-UHFFFAOYNA-N
SMILES:
CCCN(CCC)C(=O)C(CCC(O)=O)NC(=O)C1=CC=CC=C1
1
–
2
of
2
results
2
Proglumide, MedChemExpress
MedChemExpress Proglumide is a nonpeptide and orally active cholecystokinin (CCK)-A/B receptors antagonist. Proglumide selective blocks CCK’s effects in the central nervous system (CNS). Proglumide has ability to inhibit gastric secretion and to protect the gastroduodenal mucosa. Proglumide also has antiepileptic and antioxidant activities.