Learn More
Proglumide, MedChemExpress
Product Code. 16428736
Encompass_Preferred
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Product Code. 16428736
Brand: Medchem Express HYB1330
MedChemExpress Proglumide is a nonpeptide and orally active cholecystokinin (CCK)-A/B receptors antagonist. Proglumide selective blocks CCK’s effects in the central nervous system (CNS). Proglumide has ability to inhibit gastric secretion and to protect the gastroduodenal mucosa. Proglumide also has antiepileptic and antioxidant activities.
Proglumide is a nonpeptide and orally active cholecystokinin (CCK)-A/B receptors antagonist. Proglumide selective blocks CCK’s effects in the central nervous system (CNS). Proglumide has ability to inhibit gastric secretion and to protect the gastroduodenal mucosa. Proglumide also has antiepileptic and antioxidant activities.
Specifications
Chemical Name or Material Proglumide
CAS 6620-60-6
Color White
Purity Grade Notes Research
Molecular Formula C18H26N2O4
Quantity 100 mg
Solubility Information DMSO : ≥ 65 mg/mL (194.37 mM)
SMILES O=C(O)CCC(NC(C1=CC=CC=C1)=O)C(N(CCC)CCC)=O
Molecular Weight (g/mol) 334.41
Formula Weight 334.41
Percent Purity 99.65%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death