accessibility menu, dialog, popup

UserName

ClPOOO

CAS RN 6609-64-9

IUPAC Name: 2-chloro-1,3,2λ⁵-dioxaphospholan-2-one
Synonyms: 2-chloro-1,3,2λ⁵-dioxaphospholan-2-one
Molecular Weight (g/mol): 142.47
Molecular Formula: C2H4ClO3P
InChi Key: SBMUNILHNJLMBF-UHFFFAOYSA-N
SMILES: ClP1(=O)OCCO1
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
Boiling Point 
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (3)
  • (3)