accessibility menu, dialog, popup

UserName

CAS RN 66009-10-7

OOOOHOHOHOHOHHOHOOHH2O

CAS RN 66009-10-7

IUPAC Name: 2,3,4,5-tetrahydroxy-6-{[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}hexanal hydrate
Synonyms: melibiose hydrate
Molecular Weight (g/mol): 360.31
Molecular Formula: C12H24O12
InChi Key: CHIDEFLSUMQFBY-UHFFFAOYNA-N
SMILES: O.OCC1OC(OCC(O)C(O)C(O)C(O)C=O)C(O)C(O)C1O
Physical Form 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)