accessibility menu, dialog, popup

UserName

CAS RN 6600-40-4

(S)OHONH2H3C

CAS RN 6600-40-4

IUPAC Name: 2-aminopentanoic acid
Synonyms: Nva norvalines α-DL-aminopentanoic acid DL-norvaline 2-aminopentanoic acids 2-aminovaleric acids
Molecular Weight (g/mol): 117.15
Molecular Formula: C5H11NO2
InChi Key: SNDPXSYFESPGGJ-UHFFFAOYNA-N
SMILES: CCCC(N)C(O)=O
Quantity 
  • (3)
  • (1)
  • (3)
  • (1)
Percent Purity 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (2)