accessibility menu, dialog, popup

UserName

CAS RN 66-99-9

O

CAS RN 66-99-9

IUPAC Name: naphthalene-2-carbaldehyde
Synonyms: 2-naphthaldehyde
Molecular Weight (g/mol): 156.18
Molecular Formula: C11H8O
InChi Key: PJKVFARRVXDXAD-UHFFFAOYSA-N
SMILES: O=CC1=CC=C2C=CC=CC2=C1
Boiling Point 
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)