accessibility menu, dialog, popup

UserName

ONHOHN

CAS RN 66-22-8

IUPAC Name: 1,2,3,4-tetrahydropyrimidine-2,4-dione
Synonyms: uracil Ura urazil
Molecular Weight (g/mol): 112.09
Molecular Formula: C4H4N2O2
InChi Key: ISAKRJDGNUQOIC-UHFFFAOYSA-N
SMILES: O=C1NC=CC(=O)N1
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (3)
  • (3)
  • (4)
Physical Form 
  • (7)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (4)
  • (7)
  • (1)
  • (1)