accessibility menu, dialog, popup

UserName

CAS RN 6574-97-6

ClNCl

CAS RN 6574-97-6

IUPAC Name: 2,3-dichlorobenzonitrile
Synonyms: 2,3-dichlorobenzonitrile
Molecular Weight (g/mol): 172.01
Molecular Formula: C7H3Cl2N
InChi Key: OHDYZVVLNPXKDX-UHFFFAOYSA-N
SMILES: ClC1=CC=CC(C#N)=C1Cl
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results