CAS RN 65739-04-0
CAS RN 65739-04-0
IUPAC Name:
2-fluoro-N,N-dimethyl-4-nitroaniline
Synonyms:
2-fluoro-N,N-dimethyl-4-nitroaniline
Molecular Weight (g/mol):
184.17
Molecular Formula:
C8H9FN2O2
InChi Key:
QTXUWCWOGLFTTP-UHFFFAOYSA-N
SMILES:
CN(C)C1=C(F)C=C(C=C1)[N+]([O-])=O
1
–
1
of
1
results
1
2-Fluoro-N,N-dimethyl-4-nitroaniline, 97%, Thermo Scientific™
CAS: 65739-04-0 Molecular Formula: C8H9FN2O2 Molecular Weight (g/mol): 184.17 InChI Key: QTXUWCWOGLFTTP-UHFFFAOYSA-N Synonym: 2-fluoro-n,n-dimethyl-4-nitro-benzenamine,acmc-209nru,benzenamine, 2-fluoro-n,n-dimethyl-4-nitro,n,n-dimethyl-2-fluoro-4-nitroaniline PubChem CID: 23547995 IUPAC Name: 2-fluoro-N,N-dimethyl-4-nitroaniline SMILES: CN(C)C1=C(C=C(C=C1)[N+](=O)[O-])F