accessibility menu, dialog, popup

UserName

OOHNH2(S)H2NHCl

CAS RN 657-27-2

IUPAC Name: hydrogen (2S)-2,6-diaminohexanoic acid chloride
Synonyms: lysine hydrochloride
Molecular Weight (g/mol): 182.65
Molecular Formula: C6H15ClN2O2
InChi Key: BVHLGVCQOALMSV-JEDNCBNOSA-N
SMILES: [H+].[Cl-].NCCCC[C@H](N)C(O)=O
Molecular Weight (g/mol) 
  • (9)
  • (15)
Quantity 
  • (6)
  • (1)
  • (6)
  • (1)
  • (3)
  • (2)
  • (1)
  • (3)
Percent Purity 
  • (3)
  • (7)
  • (3)
  • (1)
Grade 
  • (3)
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (4)
  • (3)
  • (6)
  • (3)
  • (4)
  • (9)
  • (15)
  • (6)
  • (1)
  • (6)
  • (1)
  • (3)
  • (2)
  • (1)
  • (3)
  • (3)
  • (7)
  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (2)