accessibility menu, dialog, popup

UserName

OH(R)O(S)OH(S)OH(S)OHOHO

CAS RN 6556-12-3

IUPAC Name: (2S,3S,4S,5R)-2,3,4,5-tetrahydroxy-6-oxohexanoic acid
Synonyms: aldehydo-D-glucuronic acid D-glucuronsaeure D-glukuronsaeure
Molecular Weight (g/mol): 194.14
Molecular Formula: C6H10O7
InChi Key: IAJILQKETJEXLJ-QTBDOELSSA-N
SMILES: O[C@@H](C=O)[C@@H](O)[C@H](O)[C@H](O)C(O)=O
Percent Purity 
  • (2)
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)