accessibility menu, dialog, popup

UserName

CAS RN 65501-24-8

IUPAC Name: tripotassium 2-({2-[bis(carboxylatomethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate
Synonyms: tripotassium 2-({2-[bis(carboxylatomethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate
Molecular Weight (g/mol): 442.55
Molecular Formula: C10H17K3N2O10
InChi Key: MAPFUJCWRWFQIY-UHFFFAOYSA-K
SMILES: O.O.[K+].[K+].[K+].OC(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)