accessibility menu, dialog, popup

UserName

N

CAS RN 6542-60-5

IUPAC Name: 2-cyclopropylacetonitrile
Synonyms: 2-cyclopropylacetonitrile
Molecular Weight (g/mol): 81.12
Molecular Formula: C5H7N
InChi Key: FAUQRRGKJKMEIW-UHFFFAOYSA-N
SMILES: N#CCC1CC1
Boiling Point 
  • (3)
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results