accessibility menu, dialog, popup

UserName

CAS RN 65355-00-2

HOOH(?)

CAS RN 65355-00-2

IUPAC Name: 5H,5'H,6H,6'H,7H,7'H,8H,8'H-[1,1'-binaphthalene]-2,2'-diol
Synonyms: 5H,5'H,6H,6'H,7H,7'H,8H,8'H-[1,1'-binaphthalene]-2,2'-diol
Molecular Weight (g/mol): 294.39
Molecular Formula: C20H22O2
InChi Key: UTXIFKBYNJRJPH-UHFFFAOYSA-N
SMILES: OC1=C(C2=C(CCCC2)C=C1)C1=C(O)C=CC2=C1CCCC2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results