accessibility menu, dialog, popup

UserName

CAS RN 65330-63-4

IUPAC Name: 3-(4-hydroxyphenyl)propanehydrazide
Synonyms: 3-(4-hydroxyphenyl)propanehydrazide
Molecular Weight (g/mol): 180.21
Molecular Formula: C9H12N2O2
InChi Key: JMNHUOPRGLTNJI-UHFFFAOYSA-N
SMILES: NNC(=O)CCC1=CC=C(O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results