accessibility menu, dialog, popup

UserName

FFFOFF

CAS RN 653-37-2

IUPAC Name: 2,3,4,5,6-pentafluorobenzaldehyde
Synonyms: pentafluor-benzaldehyd
Molecular Weight (g/mol): 196.08
Molecular Formula: C7HF5O
InChi Key: QJXCFMJTJYCLFG-UHFFFAOYSA-N
SMILES: FC1=C(F)C(F)=C(C=O)C(F)=C1F
Boiling Point 
  • (6)
Molecular Weight (g/mol) 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (6)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)