CAS RN 6528-34-3
CAS RN 6528-34-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Permanent Yellow Rn, TRC
CAS: 6528-34-3 Molecular Formula: C18H18N4O6 Molecular Weight (g/mol): 386.36 IUPAC Name: 2-[(E)-(4-methoxy-2-nitrophenyl)diazenyl]-N-(2-methoxyphenyl)-3-oxobutanamide SMILES: COc1ccc(N=NC(C(=O)C)C(=O)Nc2ccccc2OC)c(c1)[N+](=O)[O-]