CAS RN 65166-97-4
CAS RN 65166-97-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(-)-Esermethole, TRC
CAS: 65166-97-4 Molecular Formula: C14H20N2O Molecular Weight (g/mol): 232.32 Synonym: (3aS,8aR)-1,2,3,3a,8,8a-Hexahydro-5-methoxy-1,3a,8-trimethylpyrrolo[2,3-b]indole,(-)-Eseroline Methyl Ether,(-)-O-Methyleseroline,NSC 340070 SMILES: COc1ccc2N(C)[C@H]3N(C)CC[C@@]3(C)c2c1