accessibility menu, dialog, popup

UserName

CAS RN 65145-13-3

OHFOHO

CAS RN 65145-13-3

IUPAC Name: 2-fluoro-4-hydroxybenzoic acid
Synonyms: 2-fluoro-4-hydroxybenzoic acid
Molecular Weight (g/mol): 156.11
Molecular Formula: C7H5FO3
InChi Key: NXWTWYULZRDBSA-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(F)C=C(O)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results