accessibility menu, dialog, popup

UserName

CAS RN 65-46-3

N(R)N(R)O(S)(R)OOHNH2HOHO

CAS RN 65-46-3

IUPAC Name: 4-amino-1-[3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2-dihydropyrimidin-2-one
Synonyms: cytidine
Molecular Weight (g/mol): 243.22
Molecular Formula: C9H13N3O5
InChi Key: UHDGCWIWMRVCDJ-UHFFFAOYNA-N
SMILES: NC1=NC(=O)N(C=C1)C1OC(CO)C(O)C1O
Quantity 
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (3)
  • (1)
Physical Form 
  • (7)
  • (4)
Percent Purity 
  • (3)
  • (3)
Grade 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (6)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (3)
  • (1)
  • (7)
  • (4)
  • (3)
  • (3)
  • (2)
  • (1)