accessibility menu, dialog, popup

UserName

H3CCH2(R)CH3O

CAS RN 6485-40-1

IUPAC Name: (5R)-2-methyl-5-(prop-1-en-2-yl)cyclohex-2-en-1-one
Synonyms: (-)-carvone (-)-p-mentha-6,8-dien-2-one (-)-(R)-carvone l-carvone (natural) levo-carvone l-carvone (R)-carvone L(-)-carvone (R)-(-)-carvone L-carvone
Molecular Weight (g/mol): 150.22
Molecular Formula: C10H14O
InChi Key: ULDHMXUKGWMISQ-SECBINFHSA-N
SMILES: CC(=C)[C@@H]1CC=C(C)C(=O)C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (6)