accessibility menu, dialog, popup

UserName

CAS RN 646477-45-4

CH3NNH2HClHCl

CAS RN 646477-45-4

IUPAC Name: 8-methyl-8-azabicyclo[3.2.1]octan-3-amine dihydrochloride
Synonyms: 8-methyl-8-azabicyclo[3.2.1]octan-3-amine dihydrochloride
Molecular Weight (g/mol): 213.15
Molecular Formula: C8H18Cl2N2
InChi Key: KVYQKWPZQKGICY-UHFFFAOYNA-N
SMILES: Cl.Cl.CN1C2CCC1CC(N)C2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results