accessibility menu, dialog, popup

UserName

CAS RN 645-92-1

NH2NONH2NHN

CAS RN 645-92-1

IUPAC Name: 4,6-diamino-2,5-dihydro-1,3,5-triazin-2-one
Synonyms: ammeline
Molecular Weight (g/mol): 127.11
Molecular Formula: C3H5N5O
InChi Key: MASBWURJQFFLOO-UHFFFAOYSA-N
SMILES: NC1=NC(=O)N=C(N)N1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)