accessibility menu, dialog, popup

UserName

ONOI

CAS RN 645-00-1

IUPAC Name: 1-iodo-3-nitrobenzene
Synonyms: 3-iodonitrobenzene
Molecular Weight (g/mol): 249.01
Molecular Formula: C6H4INO2
InChi Key: CBYAZOKPJYBCHE-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=CC(I)=C1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)