accessibility menu, dialog, popup

UserName

CAS RN 6440-02-4

H3COOSNNNH2

CAS RN 6440-02-4

IUPAC Name: ethyl 5-amino-1,2,3-thiadiazole-4-carboxylate
Synonyms: ethyl 5-amino-1,2,3-thiadiazole-4-carboxylate
Molecular Weight (g/mol): 173.19
Molecular Formula: C5H7N3O2S
InChi Key: ZUGOHJWUTNKRMG-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C(N)SN=N1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results