accessibility menu, dialog, popup

UserName

OHOHOHOHHOHO

CAS RN 643-12-9

IUPAC Name: cyclohexane-1,2,3,4,5,6-hexol
Synonyms: (-)-inositol 肌醇 inositol inositols 1,2,3,4,5,6-cyclohexanehexol
Molecular Weight (g/mol): 180.16
Molecular Formula: C6H12O6
InChi Key: CDAISMWEOUEBRE-UHFFFAOYSA-N
SMILES: OC1C(O)C(O)C(O)C(O)C1O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results