accessibility menu, dialog, popup

UserName

CAS RN 64285-95-6

FFFONCS

CAS RN 64285-95-6

IUPAC Name: 1-isothiocyanato-4-(trifluoromethoxy)benzene
Synonyms: 1-isothiocyanato-4-(trifluoromethoxy)benzene
Molecular Weight (g/mol): 219.18
Molecular Formula: C8H4F3NOS
InChi Key: JKSZUQPHKOPVHF-UHFFFAOYSA-N
SMILES: FC(F)(F)OC1=CC=C(C=C1)N=C=S
Quantity 
  • (2)
  • (1)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)