accessibility menu, dialog, popup

UserName

CAS RN 642-71-7

IUPAC Name: 3,4,5-trimethoxyphenol
Synonyms: 3,4,5-trimethoxyphenol antiarol
Molecular Weight (g/mol): 184.19
Molecular Formula: C9H12O4
InChi Key: VTCDZPUMZAZMSB-UHFFFAOYSA-N
SMILES: COC1=CC(O)=CC(OC)=C1OC
Molecular Weight (g/mol) 
  • (3)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (3)
  • (3)