accessibility menu, dialog, popup

UserName

CAS RN 642-31-9

IUPAC Name: anthracene-9-carbaldehyde
Synonyms: 9-anthracenecarboxaldehyde
Molecular Weight (g/mol): 206.24
Molecular Formula: C15H10O
InChi Key: YMNKUHIVVMFOFO-UHFFFAOYSA-N
SMILES: O=CC1=C2C=CC=CC2=CC2=CC=CC=C12
Quantity 
  • (3)
  • (3)
  • (1)
  • (2)
Percent Purity 
  • (3)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (2)
  • (3)
  • (3)
  • (3)