accessibility menu, dialog, popup

UserName

CAS RN 640769-70-6

IUPAC Name: 3-(pyrimidin-5-yl)benzaldehyde
Synonyms: 3-(pyrimidin-5-yl)benzaldehyde
Molecular Weight (g/mol): 184.2
Molecular Formula: C11H8N2O
InChi Key: RAHJVJBPRWJPHE-UHFFFAOYSA-N
SMILES: O=CC1=CC=CC(=C1)C1=CN=CN=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results