accessibility menu, dialog, popup

UserName

H3CONH(S)OHONHOOCH3CH3H3C

CAS RN 6404-26-8

IUPAC Name: 2-{[(tert-butoxy)carbonyl]amino}-6-acetamidohexanoic acid
Synonyms: 2-[(tert-butoxycarbonyl)amino]-6-acetamidohexanoic acid
Molecular Weight (g/mol): 288.34
Molecular Formula: C13H24N2O5
InChi Key: IOKOUUAPSRCSNT-UHFFFAOYNA-N
SMILES: CC(=O)NCCCCC(NC(=O)OC(C)(C)C)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results