accessibility menu, dialog, popup

UserName

CAS RN 64-02-8

NaNaNaNaOONNOOOOOO

CAS RN 64-02-8

IUPAC Name: tetrasodium 2-({2-[bis(carboxylatomethyl)amino]ethyl}(carboxylatomethyl)amino)acetate
Synonyms: Na4EDTA dequest 2016 dequest® 2016 tetrasodium edta edta, tetrasodium salt
Molecular Weight (g/mol): 380.17
Molecular Formula: C10H12N2Na4O8
InChi Key: UEUXEKPTXMALOB-UHFFFAOYSA-J
SMILES: [Na+].[Na+].[Na+].[Na+].[O-]C(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results