accessibility menu, dialog, popup

UserName

CAS RN 63932-26-3

IUPAC Name: 1-{4H,6H,7H-thieno[3,2-c]pyran-4-yl}methanamine
Synonyms: 1-{4H,6H,7H-thieno[3,2-c]pyran-4-yl}methanamine
Molecular Weight (g/mol): 169.24
Molecular Formula: C8H11NOS
InChi Key: NMVKUIFBCHQTHO-UHFFFAOYNA-N
SMILES: NCC1OCCC2=C1C=CS2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results