CAS RN 63837-33-2
CAS RN 63837-33-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
Diofenolan, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
2
Diofenolan, TRC
CAS: 63837-33-2 Molecular Formula: C18 H20 O4 Molecular Weight (g/mol): 300.35 Synonym: 1,3-Dioxolane, 2-ethyl-4-[(4-phenoxyphenoxy)methyl]-,2-Ethyl-4-[(4-phenoxyphenoxy)methyl]-1,3-dioxolane,Aware,Diofenolan IUPAC Name: 2-ethyl-4-[(4-phenoxyphenoxy)methyl]-1,3-dioxolane SMILES: CCC1OCC(COc2ccc(Oc3ccccc3)cc2)O1