CAS RN 6372-40-3
IUPAC Name:
diphenyl(propan-2-yl)phosphane
Synonyms:
isopropyldiphenylphosphane
Molecular Weight (g/mol):
228.28
Molecular Formula:
C15H17P
InChi Key:
LLZAIAIZAVMQIG-UHFFFAOYSA-N
SMILES:
CC(C)P(C1=CC=CC=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
Isopropyldiphenylphosphine, 99%
CAS: 6372-40-3 Molecular Formula: C15H17P Molecular Weight (g/mol): 228.28 MDL Number: MFCD00015028 InChI Key: LLZAIAIZAVMQIG-UHFFFAOYSA-N Synonym: isopropyldiphenylphosphine,phosphine, 1-methylethyl diphenyl,isopropyldiphenylphosphane,isopropyl diphenyl phosphine,acmc-209nho,isopropyl-diphenylphosphine,diphenyl propan-2-yl phosphane,i-propyldiphenylphosphine PubChem CID: 80754 IUPAC Name: diphenyl(propan-2-yl)phosphane SMILES: CC(C)P(C1=CC=CC=C1)C1=CC=CC=C1