accessibility menu, dialog, popup

UserName

CAS RN 637-69-4

H3COCH2

CAS RN 637-69-4

IUPAC Name: 1-ethenyl-4-methoxybenzene
Synonyms: benzene, 1-ethenyl-4-methoxy-
Molecular Weight (g/mol): 134.18
Molecular Formula: C9H10O
InChi Key: UAJRSHJHFRVGMG-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C)C=C1
Boiling Point 
  • (5)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (5)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)