accessibility menu, dialog, popup

UserName

ONOI

CAS RN 636-98-6

IUPAC Name: 1-iodo-4-nitrobenzene
Synonyms: benzene, 1-iodo-4-nitro-
Molecular Weight (g/mol): 249.01
Molecular Formula: C6H4INO2
InChi Key: SCCCFNJTCDSLCY-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(I)C=C1
Molecular Weight (g/mol) 
  • (4)
  • (5)
Quantity 
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (4)
  • (5)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)