accessibility menu, dialog, popup

UserName

CAS RN 635-80-3

IUPAC Name: 2-methylquinoline-6-carboxylic acid
Synonyms: 2-methylquinoline-6-carboxylic acid
Molecular Weight (g/mol): 187.2
Molecular Formula: C11H9NO2
InChi Key: IZONZQFTYGVOOO-UHFFFAOYSA-N
SMILES: CC1=CC=C2C=C(C=CC2=N1)C(O)=O
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)