accessibility menu, dialog, popup

UserName

CAS RN 634-97-9

HOONH

CAS RN 634-97-9

IUPAC Name: 1H-pyrrole-2-carboxylic acid
Synonyms: minalin 2-minaline PYC minaline
Molecular Weight (g/mol): 111.1
Molecular Formula: C5H5NO2
InChi Key: WRHZVMBBRYBTKZ-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CN1
Percent Purity 
  • (3)
  • (1)
  • (3)
Quantity 
  • (3)
  • (2)
  • (2)
Physical Form 
  • (3)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)