accessibility menu, dialog, popup

UserName

CAS RN 634-93-5

NH2ClClCl

CAS RN 634-93-5

IUPAC Name: 2,4,6-trichloroaniline
Synonyms: 2,4,6-trichloroaniline
Molecular Weight (g/mol): 196.46
Molecular Formula: C6H4Cl3N
InChi Key: NATVSFWWYVJTAZ-UHFFFAOYSA-N
SMILES: NC1=C(Cl)C=C(Cl)C=C1Cl
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)