accessibility menu, dialog, popup

UserName

CAS RN 634-36-6

IUPAC Name: 1,2,3-trimethoxybenzene
Synonyms: benzene, 1,2,3-trimethoxy-
Molecular Weight (g/mol): 168.19
Molecular Formula: C9H12O3
InChi Key: CRUILBNAQILVHZ-UHFFFAOYSA-N
SMILES: COC1=CC=CC(OC)=C1OC
Molecular Weight (g/mol) 
  • (5)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (5)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)