accessibility menu, dialog, popup

UserName

IH3CN

CAS RN 634-35-5

IUPAC Name: 1-ethylquinolin-1-ium iodide
Synonyms: quinolinium, 1-ethyl-, iodide
Molecular Weight (g/mol): 285.13
Molecular Formula: C11H12IN
InChi Key: PMYUGMDDIBOXQM-UHFFFAOYSA-M
SMILES: [I-].CC[N+]1=C2C=CC=CC2=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results