accessibility menu, dialog, popup

UserName

FFFSOOOFFFSOOOFFFSOOOFe

CAS RN 63295-48-7

IUPAC Name: iron(3+) tritrifluoromethanesulfonate
Synonyms: iron(3+) tritriflate
Molecular Weight (g/mol): 503.03
Molecular Formula: C3F9FeO9S3
InChi Key: OSHOQERNFGVVRH-UHFFFAOYSA-K
SMILES: [Fe+3].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results