CAS RN 63285-84-7
CAS RN 63285-84-7
IUPAC Name:
3-phenoxythiophene
Synonyms:
3-phenoxythiophene
Molecular Weight (g/mol):
176.23
Molecular Formula:
C10H8OS
InChi Key:
DXSHUWUDKNEGFS-UHFFFAOYSA-N
SMILES:
O(C1=CSC=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
3-Phenoxythiophene, 97%, Thermo Scientific™
CAS: 63285-84-7 Molecular Formula: C10H8OS Molecular Weight (g/mol): 176.23 MDL Number: MFCD00178747 InChI Key: DXSHUWUDKNEGFS-UHFFFAOYSA-N Synonym: 3-phenoxy-thiophene,acmc-20amz2 PubChem CID: 7018947 IUPAC Name: 3-phenoxythiophene SMILES: O(C1=CSC=C1)C1=CC=CC=C1